missing translation for 'onlineSavingsMsg'
Learn More

2’-O,4’-C-Methyleneuridine, MedChemExpress

Product Code. 16430817
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16430817 50 mg 50mg
16414508 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16430817 Supplier Medchem Express Supplier No. HY111639
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2’-O,4’-C-Methyleneuridine (Compound 15a) is a bicyclic nucleoside.

2’-O,4’-C-Methyleneuridine (Compound 15a) is a bicyclic nucleoside.

Specifications

Chemical Name or Material 2’-O,4’-C-Methyleneuridine
CAS 200435-92-3
Color White
Purity Grade Notes Research
Molecular Formula C10H12N2O6
Quantity 50 mg
Solubility Information DMSO : 125 mg/mL (487.88 mM; Need ultrasonic)
SMILES OC[C@@]12O[C@@H](N3C(NC(C=C3)=O)=O)[C@H](OC2)[C@@H]1O
Molecular Weight (g/mol) 256.21
Formula Weight 256.21
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.