missing translation for 'onlineSavingsMsg'
Learn More

2-O-alpha-D-Glucopyranosyl-L-ascorbic acid, 98+%

Product Code. 15448019
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448019 5 g 5g
15438019 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448019 Supplier Thermo Scientific Alfa Aesar Supplier No. J66601.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 129499-78-1 | C12H18O11 | 338.265 g/mol

2-O-alpha-D-Glucopyranosyl-L-ascorbic acid reduce the free radicals that result from UV irradiation of the skin, and significantly reduce cell damage and photo-aging. It also promotes collagen synthesis. It can function as an antioxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-O-alpha-D-Glucopyranosyl-L-ascorbic acid reduce the free radicals that result from UV irradiation of the skin, and significantly reduce cell damage and photo-aging. It also promotes collagen synthesis. It can function as an antioxidant.

Solubility
Soluble in water. (879 g/L) at 25°C.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129499-78-1
Molecular Formula C12H18O11
Molecular Weight (g/mol) 338.265
MDL Number MFCD23701380
InChI Key MLSJBGYKDYSOAE-DCWMUDTNSA-N
Synonym l-ascorbic acid 2-glucoside, aa-2g, unii-2v52r0nhxw, 2-o-alpha-d-glucopyranosyl-l-ascorbic acid, 2v52r0nhxw, l-ascorbic acid-2-glucoside, ascorbic acid 2-o-glucoside, ascorbyl glucoside, l-ascorbic acid 2-o-alpha-glucoside, l-ascorbicacid2-glucoside
PubChem CID 54693473
ChEBI CHEBI:81685
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2H-furan-5-one
SMILES C(C1C(C(C(C(O1)OC2=C(C(OC2=O)C(CO)O)O)O)O)O)O

Specifications

Melting Point 158°C to 163°C
Color White
Quantity 5 g
Solubility Information Soluble in water. (879g/L) at 25°C.
Formula Weight 338.26
Percent Purity 99.5%
Physical Form Powder
Chemical Name or Material 2-O-alpha-D-Glucopyranosyl-L-ascorbic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.