missing translation for 'onlineSavingsMsg'
Learn More

2-O-Methyl-beta-D-N-acetylneuraminic Acid Sodium Salt, TRC

Product Code. 30477669
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477669 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477669 Supplier Toronto Research Chemicals Supplier No. TRCM27540825MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-O-Methyl-beta-D-N-acetylneuraminic Acid Sodium Salt
CAS 63879-76-5
Molecular Formula C12H20NNaO9
Quantity 25 mg
Synonym 2-O-Methyl-N-acetyl-β-neuraminic Acid, N-Acetyl-β-neuraminic Methyl Glycoside Sodium Salt
InChI Formula InChI=1S/C12H21NO9.Na/c1-5(15)13-8-6(16)3-12(21-2,11(19)20)22-10(8)9(18)7(17)4-14;/h6-10,14,16-18H,3-4H2,1-2H3,(H,13,15)(H,19,20);/q;+1/p-1/t6-,7+,8+,9+,10+,12-;/m0./s1
SMILES O=C([C@]1(O[C@@]([C@@H]([C@H](C1)O)NC(C)=O)([H])[C@H](O)[C@H](O)CO)OC)O[Na]
IUPAC Name sodium (2S,4S,5R,6R)-5-acetamido-4-hydroxy-2-methoxy-6-((1R,2R)-1,2,3-trihydroxypropyl)tetrahydro-2H-pyran-2-carboxylate
Molecular Weight (g/mol) 345.28
Formula Weight 345.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.