missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Octanol, 98%

Product Code. 11428826
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
This item is not returnable. View return policy

Product Code. 11428826

Brand: Thermo Scientific Alfa Aesar A16985.0F

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-96-6 | C8H18O | 130.231 g/mol

(±)-2-Octanol is used in flavors, paints and coatings, defoaming agent, low-volatility solvent, lubricants, fuels, paints and coatings. It is used as a frother in mineral floatation and wetting agent. It acts as an intermediate in the preparation of surfactants, cosmetic emollients esters and plasticizers. Further, it is also used as a reagent in the synthesis of pharmaceutical compounds and piperine derivatives. In addition to this, it is considered as an alternative of 2-ethylhexanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(^+)-2-Octanol is used in flavors, paints and coatings, defoaming agent, low-volatility solvent, lubricants, fuels, paints and coatings. It is used as a frother in mineral floatation and wetting agent. It acts as an intermediate in the preparation of surfactants, cosmetic emollients esters and plasticizers. Further, it is also used as a reagent in the synthesis of pharmaceutical compounds and piperine derivatives. In addition to this, it is considered as an alternative of 2-ethylhexanol.

Solubility
Miscible with diethyl ether, acetone, alcohol, most common organic solvents, aromatic and aliphatic hydrocarbons. Slightly miscible with cold water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-96-6
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00004591
InChI Key SJWFXCIHNDVPSH-UHFFFAOYSA-N
Synonym 2-octanol, 2-octyl alcohol, hexylmethylcarbinol, 2-hydroxyoctane, sec-caprylic alcohol, 1-methylheptanol, methylhexylcarbinol, 1-methylheptyl alcohol, dl-2-octanol, n-octan-2-ol
PubChem CID 20083
ChEBI CHEBI:37869
IUPAC Name octan-2-ol
SMILES CCCCCCC(C)O

Specifications

Melting Point -38°C
Density 0.819
Boiling Point 179°C to 180°C
Flash Point 76°C (169°F)
Odor Pleasant
Refractive Index 1.426
Quantity 2500 mL
Beilstein 1719325
Merck Index 14,6752
Solubility Information Miscible with diethyl ether,acetone,alcohol,most common organic solvents,aromatic and aliphatic hydrocarbons. Slightly miscible with cold water.
Formula Weight 130.23
Percent Purity 98%
Chemical Name or Material (+/-)-2-Octanol
Show More Show Less
compliance-icons
Product Identifier
  • (+/-)-2-Octanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 2-Octanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.