missing translation for 'onlineSavingsMsg'
Learn More

2-Octanone oxime, 99%, Thermo Scientific™

Product Code. 10419543 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10419543 25 g Glass bottle 25g
10772475 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10419543 Supplier Thermo Scientific Acros Supplier No. 390280250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7207-49-0
Molecular Formula C8H17NO
Molecular Weight (g/mol) 143.23
MDL Number MFCD00089167
InChI Key GZRPVYSKBVDCBV-HJWRWDBZSA-N
Synonym 2-octanone, oxime, 2-octanone oxime, n-octan-2-ylidene hydroxylamine, 2-hydroxyiminooctane, z-n-octan-2-ylidene hydroxylamine
PubChem CID 9562584
IUPAC Name (NE)-N-octan-2-ylidenehydroxylamine
SMILES CCCCCC\C(C)=N/O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8800g/mL
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)5C(CH3)=NOH
Refractive Index 1.4510 to 1.453
Quantity 25 g
Specific Gravity 0.88
Formula Weight 143.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Octanone oxime
Show More Show Less
compliance-icons
Product Identifier
  • 2-Octanone oxime
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.