missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2'-OMe-Ac-C Phosphoramidite, standard grade, serum vial bottle

Product Code. 18044080 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18044080 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18044080 Supplier Thermo Scientific™ Supplier No. 27182302
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Name: 5'-O-Dimethoxytrityl-N4-acetyl-2'-O-methyl-cytidine-3'-O-(2-cyanoethyl-N,N-diisopropyl) phosphoramidite

Thermo Scientific RNA phosphoramidites ensure a high level of quality and reliability for RNA oligonucleotide synthesis. Our phosphoramidites are manufactured to an exacting standard to reduce levels of critical impurities.

Highlights

  • RNA oligonucleotides synthesized using 2'-Fluoro U phosphoramidites have increased thermal stability of RNA duplexes and nuclease resistance of single-stranded RNA
  • These phosphoramidites can be incorporated into custom siRNA oligonucleotides for use in gene-targeting and suppression studies
  • Rigorously controlled manufacturing processes minimize levels of reactive P(III) impurities which could be incorporated into the oligonucleotide product and/or result in the accumulation of by-products which can be difficult to remove during purification
  • NMR analysis is conducted on each lot to assure that the levels of critical impurities are within specifications

Process and Supply Chain Control

  • Processes are strictly controlled and monitored from material procurement through final QC and product release under ISO9001 standards, providing batch-to-batch reproducibility
  • All standard RNA phosphoramidites are supplied with lot-specific quality control data including HPLC and phosphorous NMR analyses

Flexible Packaging

  • Common pack sizes fit most commercially available synthesizers and are in stock for immediate delivery
  • Custom and bulk packaging are available
TRUSTED_SUSTAINABILITY

Specifications

Format Powder
For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type 2'-OMe-Ac-C CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Cytidine (C)
Reagent Features 2'-OMe modified, Fast deprotection
Shipping Condition Room Temperature
Purity 98%
Quantity 1 g
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.