missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2'-OMe-iPrPAC-G Phosphoramidite, standard grade, 28-400 finish bottle

Product Code. 17967142 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17967142 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17967142 Supplier Thermo Scientific™ Supplier No. 27182611
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

2'-OMe-iPrPAC-G CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with 2'-O-methyl protection, for use in oligonucleotide synthesis.

2'-OMe-iPrPAC-G CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with 2'-O-methyl protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methyl (OMe) oligo modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The 'standard grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 30-mL amber bottle with a 28-400 finish neck style, which is compatible with most MerMade synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N2-isopropylphenoxyacetyl-2'-O-methyl-guanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: C52H62N7O10P
Base: G
Base Protection: iPrPAC
Formula weight: 976.1 g/mol
HPLC purity: ≥98.0%
31P NMR purity: ≥98.0%

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) MerMade Synthesizers
Product Type 2in-OMe-iPrPAC-G CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Guanosine (G)
Reagent Features 2in-OMe modified, Fast deprotection
Shipping Condition Room Temperature
Purity 98%
Quantity 1 g
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type RNA
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.