Learn More
Thermo Scientific™ 2'-OMe-PAC-A Phosphoramidite, standard grade, 28-400 finish bottle
Shop All Thermo Scientific ProductsDescription
2'-OMe-PAC-A CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with 2'-O-methyl protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methyl (OMe) oligo modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The 'standard grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 30-mL amber bottle with a 28-400 finish neck style, which is compatible with most MerMade synthesizers.
Features:
• Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N6-phenoxyacetyl-2'-O-methyl-adenosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C49H56N7O9P
• Base: A
• Base Protection: PAC
• Formula weight: 918.0 g/mol
• CAS number: 128219-82-9
• HPLC purity: ≥98.0%
• 31P NMR purity: ≥98.0%
Specifications
Specifications
| For Use With (Equipment) | MerMade Synthesizers |
| Product Type | 2in-OMe-PAC-A CEP |
| Content And Storage | Store in freezer (at or below -20°C). |
| Nucleoside Type | Adenosine (A) |
| Reagent Features | 2in-OMe modified, Fast deprotection |
| Shipping Condition | Room Temperature |
| Purity | 98% |
| Quantity | 1 g |
| Reagent Type | Phosphoramidite |
| Grade | Standard |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.