missing translation for 'onlineSavingsMsg'
Learn More

2-Oxa-6-azaspiro[3.3]heptane hemioxalate, 96%, Thermo Scientific™

Product Code. 15464078
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15464078 500 mg 500mg
15454078 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15464078 Supplier Thermo Scientific Alfa Aesar Supplier No. H57634.ME
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1045709-32-7
Molecular Formula C12H20N2O6
Molecular Weight (g/mol) 288.30
MDL Number MFCD16652332
InChI Key RXBYDYSXIHJXNO-UHFFFAOYSA-N
PubChem CID 54594442
SMILES OC(=O)C(O)=O.C1NCC11COC1.C1NCC11COC1

Specifications

Quantity 500 mg
IUPAC Name bis(2-oxa-6-azaspiro[3.3]heptane); oxalic acid
Formula Weight 144.15
Percent Purity 96%
Chemical Name or Material 2-Oxa-6-azaspiro[3.3]heptane hemioxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.