missing translation for 'onlineSavingsMsg'
Learn More

2-Oxa-6-azaspiro[3.4]octane, 95%

Product Code. 15414038
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15414038 250 mg 250mg
15424038 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15414038 Supplier Thermo Scientific Alfa Aesar Supplier No. H57579.MD
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 220290-68-6 | C6H11NO | 113.16 g/mol

2-Oxa-6-azaspiro[3.4]octane is involved in the preparation of azaspirocycle or azetidine substituted 4-anilinoquinazoline derivatives, which exhibits epidermal growth factor receptor (EGFR) inhibitory activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Oxa-6-azaspiro[3.4]octane is involved in the preparation of azaspirocycle or azetidine substituted 4-anilinoquinazoline derivatives, which exhibits epidermal growth factor receptor (EGFR) inhibitory activities.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids, reducing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220290-68-6
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD14586455
InChI Key ZHAIMJRKJKQNQI-UHFFFAOYSA-N
Synonym 2-oxa-6-azaspiro 3.4 octane, 2-oxa-6-aza-spiro 3.4 octane, 2-oxa-6-azaspiro 3,4 octane, acmc-209fqv, 2-oxa-7-azaspiro 3.4 octane, 2-oxa-6-aza-spiro 3,4 octane, spiro pyrrolidine-3,3'-oxetane
PubChem CID 53438323
IUPAC Name 2-oxa-7-azaspiro[3.4]octane
SMILES C1CNCC12COC2

Specifications

Density 1.08
Boiling Point 187°C
Flash Point 63°C (145°F)
Quantity 250 mg
Solubility Information Soluble in water.
Formula Weight 113.16
Percent Purity 95%
Chemical Name or Material 2-Oxa-6-azaspiro[3.4]octane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.