missing translation for 'onlineSavingsMsg'
Learn More

2-Oxa-7-azaspiro[3.5]nonane hemioxalate, 97%

Product Code. 15404248
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15404248 250 mg 250mg
15414248 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15404248 Supplier Thermo Scientific Alfa Aesar Supplier No. H57834.MD
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1429056-28-9 | C16H28N2O6 | 344.408 g/mol

Spiro forms of oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photo chromic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Spiro forms of oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photo chromic materials.

Notes
Hygroscopic. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1429056-28-9
Molecular Formula C16H28N2O6
Molecular Weight (g/mol) 344.408
MDL Number MFCD11035888
InChI Key WWVUFRRXXSVWBJ-UHFFFAOYSA-N
Synonym 2-oxa-7-azaspiro 3.5 nonane oxalate 2:1, 2-oxa-7-azaspiro 3.5 nonane hemioxalate, bis 2-oxa-7-azaspiro 3.5 nonane ; oxalic acid, 2-oxa-7-azaspiro 3.5 nonan hemioxalate, 2-oxa-7-azaspiro 3,5 nonane hemioxalate, bis 2-oxa-7-azaspiro 3.5 nonane oxalate, 2-oxa-7-azaspiro 3.5 nonane hemi-oxalate
PubChem CID 71463619
IUPAC Name 2-oxa-7-azaspiro[3.5]nonane;oxalic acid
SMILES C1CNCCC12COC2.C1CNCCC12COC2.C(=O)(C(=O)O)O

Specifications

Melting Point 208°C
Color White to Gray
Density 1.048 g/mL
Quantity 250 mg
Formula Weight 172.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Oxa-7-azaspiro[3.5]nonane hemioxalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.