missing translation for 'onlineSavingsMsg'
Learn More

2-Oxazolidinone, 99%

Product Code. 11431918
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431918 25 g 25g
11441918 100 g 100g
11451918 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431918 Supplier Thermo Scientific Alfa Aesar Supplier No. A14543.14
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 497-25-6 | C3H5NO2 | 87.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Oxazolidinone is used as antimicrobials, in organisms it act as a protein synthesis inhibitors, targeting an early step involving the binding of N-formylmethionyl-tRNA to the ribosome. They are used for chiral synthesis. It has been used as an ethanolamine equivalent, also behaves as an aziridine equivalent as in the conversion of thiophenols or amines to their aminoethyl derivatives.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-25-6
Molecular Formula C3H5NO2
Molecular Weight (g/mol) 87.08
MDL Number MFCD00005268
InChI Key IZXIZTKNFFYFOF-UHFFFAOYSA-N
Synonym 2-oxazolidone, oxazolidin-2-one, 2-oxazolidinone, oxazolidinone, oxazolidone, oxazolidinones, unii-z4d49w92pp, carbamic acid, 2-hydroxyethyl-, gamma-lactone, wln: t5mvotj, carbamic acid, .gamma.-lactone
PubChem CID 73949
ChEBI CHEBI:1237
IUPAC Name 1,3-oxazolidin-2-one
SMILES O=C1NCCO1

Specifications

Melting Point 84°C to 89°C
Boiling Point 220°C (48 mmHg)
Quantity 25 g
Beilstein 106251
Solubility Information Soluble in water.
Formula Weight 87.08
Percent Purity 99%
Chemical Name or Material 2-Oxazolidinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.