missing translation for 'onlineSavingsMsg'
Learn More

2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonyl chloride, 95%, Thermo Scientific™

Product Code. 10245994
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10245994 1 g Amber Glass Bottle 1g
10295704 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10245994 Supplier Thermo Scientific Maybridge Supplier No. CC75703DA
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 27685-90-1
Molecular Formula C7H4ClNO4S
Molecular Weight (g/mol) 233.62
MDL Number MFCD03001119
InChI Key IBHCNMJSVDRBSY-UHFFFAOYSA-N
Synonym 2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonyl chloride, 2-oxo-2,3-dihydrobenzo d oxazole-6-sulfonyl chloride, 6-benzoxazolesulfonyl chloride, 2,3-dihydro-2-oxo, 2-oxo-2,3-dihydro-benzooxazole-6-sulfonyl chloride, benzoxazalone, 6-sulfochloride, 2,3-dihydro-2-oxobenzoxazole-6-sulphonyl chloride, 2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonylchloride, 6-chlorosulfonyl-3-hydrobenzoxazol-2-one, pubchem5677, acmc-209gyq
PubChem CID 119698
IUPAC Name 2-oxo-3H-1,3-benzoxazole-6-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C2NC(=O)OC2=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 233.63
Percent Purity 95%
Chemical Name or Material 2-Oxo-2, 3-dihydro-1, 3-benzoxazole-6-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.