missing translation for 'onlineSavingsMsg'
Learn More

2-Oxo-3-hydroxy-N-methyl-N-propyl D-Lysergamide(Mixture of Diastereomers), TRC

Product Code. 30427387
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30427387 10 mg
30409341 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30427387 Supplier Toronto Research Chemicals Supplier No. TRCO85688010MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 249921-57-1
Molecular Formula C20H25N3O3
Molecular Weight (g/mol) 355.44
Synonym (8beta)-9,10-Didehydro-2,3-dihydro-3-hydroxy-N,6-dimethyl-2-oxo-N-propylergoline-8-carboxamide, 2-Oxo-3-hydroxy LAMPA, 2-Oxo-3-hydroxylysergic Acid Methyl Propyl Amide
IUPAC Name (6aR,9R)-5a-hydroxy-N,7-dimethyl-5-oxo-N-propyl-6,6a,8,9-tetrahydro-4H-indolo[4,3-fg]quinoline-9-carboxamide
SMILES CCCN(C)C(=O)[C@H]1CN(C)[C@@H]2CC3(O)C(=O)Nc4cccc(C2=C1)c34

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H25N3O3/c1-4-8-22(2)18(24)12-9-14-13-6-5-7-15-17(13)20(26,19(25)21-15)10-16(14)23(3)11-12/h5-7,9,12,16,26H,4,8,10-11H2,1-3H3,(H,21,25)/t12-,16-,20?/m1/s1
Formula Weight 355.1896
Chemical Name or Material 2-Oxo-3-hydroxy-N-methyl-N-propyl D-Lysergamide(Mixture of Diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.