missing translation for 'onlineSavingsMsg'
Learn More

2-Pentanone, 99%, pure

Product Code. 10606264 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10606264 2.5 L Glass bottle 2.5 L
10193450 250 mL Glass bottle 250mL
10034143 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10606264 Supplier Thermo Scientific Acros Supplier No. 123500025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-87-9 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-87-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
InChI Key XNLICIUVMPYHGG-UHFFFAOYSA-N
Synonym 2-pentanone, methyl propyl ketone, ethyl acetone, pentanone, ethylacetone, propyl methyl ketone, metylopropyloketon, methyl n-propyl ketone, methylpropyl ketone, methyl-propyl-cetone
PubChem CID 7895
ChEBI CHEBI:16472
IUPAC Name pentan-2-one
SMILES CCCC(=O)C

Specifications

Melting Point -78.0°C
Density 0.8090g/mL
Boiling Point 100.0°C to 103.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO(CH2)CH3
Refractive Index 1.3890 to 1.3910
Quantity 2.5 L
Beilstein 01,676
Merck Index 15,6187
Specific Gravity 0.809
Solubility Information Solubility in water: 43g/L (20°C). Other solubilities: soluble in alcohol and ether
Viscosity 0.5 mPa.s (20°C)
Formula Weight 86.13
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material 2-Pentanone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Pentanone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Methyl propyl ketone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.