missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pentanone, CHROMASOLV™, for HPLC, 99.5%, Honeywell Riedel-de Haën™
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| CAS | 107-87-9 |
| Absorbance | max. 0.005 at 400 nm, max. 0.005 at 370 nm, max. 0.01 at 350 nm, max. 0.10 at 340 nm, max. 1.0 at 330 nm |
| CAS Max % | ≤100.0000% |
| Melting Point | -78°C (literature) |
| Density | 0.809 g/mL at 25 °C(lit.) |
| Boiling Point | 101°C to 105°C |
| Flash Point | 7°C |
| Packaging | Glass Bottle |
| Molecular Formula | C5H10O |
| Linear Formula | CH3COCH2CH2CH3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?