missing translation for 'onlineSavingsMsg'
Learn More

2-Pentanone, TRC

Product Code. 30421937
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. Quantity unitSize
30421937 500 g 500g
1 options
This item is not returnable. View return policy

Product Code. 30421937

Brand: Toronto Research Chemicals TRCP274020500G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methylpropylketone
CAS 107-87-9
Molecular Formula C5 H10 O
Quantity 500 g
Synonym 2-Pentanone, 4-Methyl-2-butanone, Ethylacetone, Methyl n-propyl ketone, Methyl propyl ketone, NSC 5350, Propyl methyl ketone
InChI Formula InChI=1S/C5H10O/c1-3-4-5(2)6/h3-4H2,1-2H3
SMILES CCCC(=O)C
IUPAC Name pentan-2-one
Molecular Weight (g/mol) 86.13
Formula Weight 86.0732
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.