missing translation for 'onlineSavingsMsg'
Learn More

2-Pentynoic acid, 97%

Product Code. 15490188
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490188 1 g 1g
15400198 5 g 5g
15410198 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490188 Supplier Thermo Scientific Alfa Aesar Supplier No. H53411.03
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5963-77-9 | C5H6O2 | 98.101 g/mol

2-Pentynoic acid is used in the stereoselective synthesis of vinylstannanes. It is also used in the synthesis of spiroindolines derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pentynoic acid is used in the stereoselective synthesis of vinylstannanes. It is also used in the synthesis of spiroindolines derivatives.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5963-77-9
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.101
MDL Number MFCD00014370
InChI Key MINRDQDGBLQBGD-UHFFFAOYSA-N
Synonym 2-pentynoic acid, pentynoic acid, 2-pentynoicacid, 2-pentyneoic acid, acmc-1ay8s
PubChem CID 324379
IUPAC Name pent-2-ynoic acid
SMILES CCC#CC(=O)O

Specifications

Melting Point 47°C to 53°C
Flash Point 96°C (205°F)
Linear Formula CH3CH2CH≡CCO2H
Quantity 1 g
Formula Weight 98.1
Percent Purity 97%
Chemical Name or Material 2-Pentynoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.