missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethanol, TRC

Product Code. 17951555
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17951555 25 g 25g
17944395 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17951555 Supplier Toronto Research Chemicals Supplier No. TRCP29790025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-99-6
Molecular Formula C8 H10 O2
Molecular Weight (g/mol) 138.16
Synonym Phenoxyethanol, (2-Hydroxyethoxy)benzene, β-Hydroxyethyl phenyl ether, β-Phenoxyethanol, β-Phenoxyethyl alcohol, 1-Hydroxy-2-phenoxyethane, 2-Hydroxyethyl phenyl ether, 2-Phenoxyethanol, 2-Phenoxyethyl alcohol, Arosol, Dalpad A, Dowanol EP, Dowanol EPh, Emeressence 1160, Ethylene glycol monophenyl ether, Ethylene glycol phenyl ether, H 4644, Hisolve EPH, NSC 1864, Newpol EFP, PHE, PHE (alcohol), PHE-G, PHE-S, Phenova, Phenoxethol, Phenoxetol, Phenoxyethyl alcohol, Phenyl cellosolve, Plastilit DS 3431, Rokafenol F 1, Sepicide LD
IUPAC Name 2-phenoxyethanol
SMILES OCCOc1ccccc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H10O2/c9-6-7-10-8-4-2-1-3-5-8/h1-5,9H,6-7H2
Formula Weight 138.0681
Percent Purity >95%
Chemical Name or Material Phenoxyethanol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.