missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethyl acrylate, tech. 85%, stab. with 4-methoxyphenol, Thermo Scientific Chemicals

Product Code. 15450827
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15450827 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15450827 Supplier Thermo Scientific Alfa Aesar Supplier No. L12120.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 48145-04-6
Density 1.104
Boiling Point 103°C to 104°C (1 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C11H12O3
Linear Formula C6H5O(CH2)2O2CCH=CH2
Refractive Index 1.519
MDL Number MFCD00053696
Quantity 50 g
UN Number UN3082
Beilstein 2102773
Synonym 2-phenoxyethyl acrylate, phenoxyethyl acrylate, light ester po-a, chemlink 160, ebecryl 110, 2-propenoic acid, 2-phenoxyethyl ester, ethylene glycol phenyl ether acrylate, 2-phenoxyethanol acrylate, phenyl cellosolve acrylate, acrylic acid, 2-phenoxyethyl ester
InChI Key RZVINYQDSSQUKO-UHFFFAOYSA-N
SMILES C=CC(=O)OCCOC1=CC=CC=C1
IUPAC Name 2-phenoxyethyl prop-2-enoate
Molecular Weight (g/mol) 192.21
PubChem CID 39485
Formula Weight 192.22
Percent Purity 85%
Grade Technical
Chemical Name or Material 2-Phenoxyethyl acrylate, stabilized with 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.