missing translation for 'onlineSavingsMsg'
Learn More

2-(Phenoxymethyl)oxirane, TRC

Product Code. 30416799
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416799 50 g 50g
30421611 10 g -
30418523 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30416799 Supplier Toronto Research Chemicals Supplier No. TRCP31861550G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-60-1
Molecular Formula C9 H10 O2
Molecular Weight (g/mol) 150.174
Synonym Oxirane, 2-(phenoxymethyl)-, Oxirane, (phenoxymethyl)- (9CI), Propane, 1,2-epoxy-3-phenoxy- (8CI), 2-(Phenoxymethyl)oxirane, (Phenoxymethyl)oxirane, (Phenyloxymethyl)oxirane, (±)-(Phenoxymethyl)oxirane, (±)-1,2-Epoxy-3-phenoxypropane, (±)-Glycidyl phenyl ether, (±)-Phenyl glycidyl ether, 1,2-Epoxy-3-phenoxypropane, 1-Phenoxy-2,3-epoxypropane, 2,3-Epoxy-1-phenoxypropane, 2,3-Epoxypropoxybenzene, 2,3-Epoxypropyl phenyl ether, 3-(Phenoxy)propylene epoxide, 3-(Phenyloxy)-1,2-epoxypropane, 3-Phenoxy-1,2-epoxypropane, 3-Phenoxy-1,2-propylene oxide, 3-Phenoxy-1-propene oxide, 3-Phenoxypropylene oxide, Denacol EX 141, Diluent 690, EX 141, Epiol P, Epoxy 690, Glycidol phenyl ether, Glycidyl phenyl ether, Heloxy 63, Heloxy Modifier 63, JX 016, K 103, NSC 53476, NSC 635, O-Phenylglycidol, Oxiranylmethyl phenyl ether, PGE-H, Phenol glycidyl ether, Phenyl 2,3-epoxypropyl ether, Phenyl glycidyl ether, YS 690, γ-Phenoxypropylene oxide
IUPAC Name 2-(phenoxymethyl)oxirane
SMILES C(Oc1ccccc1)C2CO2

Specifications

Quantity 50 g
InChI Formula InChI=1S/C9H10O2/c1-2-4-8(5-3-1)10-6-9-7-11-9/h1-5,9H,6-7H2
Formula Weight 150.0681
Chemical Name or Material 2-(Phenoxymethyl)oxirane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.