missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxypropylamine, 98%

Product Code. 11464630
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464630 1 g 1g
11454630 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464630 Supplier Thermo Scientific Alfa Aesar Supplier No. H50720.03
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6437-49-6 | C9H14ClNO | 187.667 g/mol

2-Phenoxypropylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenoxypropylamine is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6437-49-6
Molecular Formula C9H14ClNO
Molecular Weight (g/mol) 187.667
MDL Number MFCD07781046
InChI Key CGOBBGUREQJPPH-UHFFFAOYSA-N
Synonym 2-phenoxypropylamine hydrochloride, 2-phenoxypropan-1-amine hydrochloride, 2-phenoxypropylamine hcl, 1-aminopropan-2-yl oxy benzene hydrochloride, acmc-20annz, 2-phenoxypropan-1-amine-hydrogen chloride 1/1
PubChem CID 24212021
IUPAC Name 2-phenoxypropan-1-amine;hydrochloride
SMILES CC(CN)OC1=CC=CC=C1.Cl

Specifications

Quantity 1 g
UN Number UN3259
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 151.21
Percent Purity 98%
Chemical Name or Material 2-Phenoxypropylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.