missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-1,3-propanediol, 98%, Thermo Scientific™

Product Code. 15555176 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15555176 25 g 25g
15535176 1 g 1g
15545176 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15555176 Supplier Thermo Scientific Acros Supplier No. 455900250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1570-95-2
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
InChI Key BPBDZXFJDMJLIB-UHFFFAOYSA-N
Synonym 2-phenyl-1,3-propanediol, 2-phenyl-1,3-propanediole, 2-phenyl-propane-1,3-diol, 2-phenyl-1,3-propane diol, unii-9f93674jbb, 1,3-propanediol, 2-phenyl, phenylpropanediol, pubchem20386, rarechem al bd 1370, acmc-1bz5a
PubChem CID 254178
IUPAC Name 2-phenylpropane-1,3-diol
SMILES C1=CC=C(C=C1)C(CO)CO

Specifications

Melting Point 54.0°C to 56.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 2-Phenyl-1,3-propanediol
compliance-icons
Product Identifier
  • 2-Phenyl-1,3-propanediol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.