missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-1,3-thiazole-4-carbaldehyde, ≥90%, Thermo Scientific™

Product Code. 10059211
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10059211 1 g Amber Glass Bottle 1g
10049211 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10059211 Supplier Thermo Scientific Maybridge Supplier No. CC18304DA
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 20949-81-9
Molecular Formula C10H7NOS
Molecular Weight (g/mol) 189.232
MDL Number MFCD02681934
InChI Key OLLKCCGWRITPOV-UHFFFAOYSA-N
PubChem CID 736524
IUPAC Name 2-phenyl-1,3-thiazole-4-carbaldehyde
SMILES C1=CC=C(C=C1)C2=NC(=CS2)C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2-phenylthiazole-4-carbaldehyde, 4-thiazolecarboxaldehyde, 2-phenyl, 2-phenyl-4-formylthiazole, 4-formyl-2-phenyl-1,3-thiazole, 4-thiazolecarboxaldehyde,2-phenyl, 2-phenyl-thiazole-4-carbaldehyde, 2-phenyl-4-thiazolecarboxaldehyde, 4-formyl-1,3-thiazol-2-yl benzene, 2-phenyl-1,3-thiazole-4-carboxaldehyde
Formula Weight 189.24
Percent Purity 95%
Chemical Name or Material 2-Phenyl-1, 3-thiazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.