missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Phenyl-1-propanol, 97%

Product Code. 11485467
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485467 50 g 50g
11495467 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485467 Supplier Thermo Scientific Alfa Aesar Supplier No. B23473.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1123-85-9 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1123-85-9
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00004736
InChI Key RNDNSYIPLPAXAZ-UHFFFAOYSA-N
Synonym 2-phenyl-1-propanol, hydratropic alcohol, hydratropyl alcohol, 2-phenylpropanol-1, beta-methylphenethyl alcohol, 2-phenylpropyl alcohol, 1-propanol, 2-phenyl, 1-hydroxy-2-phenylpropane, 2-phenylpropanol, benzeneethanol, .beta.-methyl
PubChem CID 14295
IUPAC Name 2-phenylpropan-1-ol
SMILES CC(CO)C1=CC=CC=C1

Specifications

Melting Point -37°C
Density 0.975
Boiling Point 220°C
Flash Point 108°C (226°F)
Refractive Index 1.5262
Quantity 50 g
Beilstein 1906760
Formula Weight 136.19
Percent Purity 97%
Chemical Name or Material (+/-)-2-Phenyl-1-propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.