missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-imidazoline, 98+%, Thermo Scientific™

Product Code. 10773391 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
250g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10773391 5 g Glass bottle 5g
10773581 100 g Glass bottle 100g
10591641 250 g Glass bottle 250g
10674712 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10773391 Supplier Thermo Scientific Acros Supplier No. 199730050
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 936-49-2
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD00005180
InChI Key BKCCAYLNRIRKDJ-UHFFFAOYSA-N
Synonym 2-phenyl-2-imidazoline, 2-phenylimidazoline, 1h-imidazole, 4,5-dihydro-2-phenyl, 2-imidazoline, 2-phenyl, 4,5-dihydro-2-phenyl-1h-imidazole, 2-phenyl imidazoline, 2-phenyl-2-imidazolin, acmc-209rmt, labotest-bb lt00000058, 5-23-06-00425 beilstein handbook reference
PubChem CID 13639
IUPAC Name 2-phenyl-4,5-dihydro-1H-imidazole
SMILES C1CN=C(N1)C1=CC=CC=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 94°C to 99°C
Color Yellow
Boiling Point 188°C (20.0 mmHg)
Flash Point 201°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 5 g
Beilstein 23, V,6, 425
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols
Formula Weight 146.19
Percent Purity 98+%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 2-Phenyl-2-imidazoline, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Phenyl-2-imidazoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.