missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-piperidinoacetonitrile, 97%, Thermo Scientific™

Product Code. 10651565
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10651565 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10651565 Supplier Thermo Scientific Maybridge Supplier No. XBX00107DA
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5766-79-0
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C13H16N2
MDL Number MFCD00023711
Quantity 1 g
Synonym 2-phenyl-2-piperidinoacetonitrile, usaf a-2217, 2-phenyl-2-piperidin-1-yl acetonitrile, acetonitrile, 2-phenyl-2-piperidino, 1-piperidineacetonitrile, alpha-phenyl, 1-piperidineacetonitrile, .alpha.-phenyl, alpha-tolunitrile-alpha-piperidino, alpha-tolunitrile, alpha-piperidino, alpha-1-piperidyl-alpha-tolunitrile, 1-a-cyanobenzyl-piperidine
InChI Key IXQJQHORYHYKBD-UHFFFAOYSA-N
SMILES C1CCN(CC1)C(C#N)C2=CC=CC=C2
IUPAC Name 2-phenyl-2-piperidin-1-ylacetonitrile
Molecular Weight (g/mol) 200.285
PubChem CID 21989
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material 2-Phenyl-2-piperidinoacetonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.