missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-3-butyn-2-ol, 98%

Product Code. 15555056 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15555056 25 g 25g
12574276 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15555056 Supplier Thermo Scientific Acros Supplier No. 199690250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 127-66-2 | C10H10O | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-66-2
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
InChI Key KSLSOBUAIFEGLT-UHFFFAOYSA-N
Synonym 2-phenyl-3-butyn-2-ol, methylphenylethynylcarbinol, 3-hydroxy-3-phenyl-1-butyne, 3-butyn-2-ol, 2-phenyl, 3-phenyl-butin-1-ol-3, alpha-ethynyl-alpha-methylbenzyl alcohol, alpha-ethynyl-alpha-methylbenzenemethanol, benzenemethanol, .alpha.-ethynyl-.alpha.-methyl, 3-phenyl-butin-1-ol-3 german, benzenemethanol, alpha-ethynyl-alpha-methyl
PubChem CID 92098
IUPAC Name 2-phenylbut-3-yn-2-ol
SMILES CC(C#C)(C1=CC=CC=C1)O

Specifications

Melting Point 50.0°C to 52.0°C
Boiling Point 102.0°C to 103.0°C (12.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CC(C6H5)(OH)CH3
Quantity 25 g
Beilstein 06,II,559
Merck Index 10,7158
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetone, benzene and most other common, organic solvents
Formula Weight 146.19
Percent Purity 98%
Chemical Name or Material 2-Phenyl-3-butyn-2-ol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Phenyl-3-butyn-2-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.