missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-5-oxazolone, 97%

Product Code. 11352705
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352705 25 g 25g
11332705 1 g 1g
11342705 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11352705 Supplier Thermo Scientific Alfa Aesar Supplier No. L00194.14
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1199-01-5 | C9H7NO2 | 161.16 g/mol

2-Phenyl-5-oxazolone is a reagent used as a masked glycine equivalent and for the formation of heterocyclic structures, it can be used to produce 4-benzylidene-2-phenyl-4H-oxazol-5-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenyl-5-oxazolone is a reagent used as a masked glycine equivalent and for the formation of heterocyclic structures, it can be used to produce 4-benzylidene-2-phenyl-4H-oxazol-5-one.

Solubility
Soluble in most common organic solvents.

Notes
Air Sensitive. Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1199-01-5
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00014517
InChI Key QKCKCXFWENOGER-UHFFFAOYSA-N
Synonym 2-phenyloxazol-5 4h-one, phenyloxazolone, 2-phenyl-5-oxazolone, 2-phenyloxazolone, 2-phenyloxazolin-5-one, 5 4h-oxazolone, 2-phenyl, 2-phenyl-5 4h-oxazolone, 2-phenyl-1,3-oxazol-5 4h-one, 2-phenyl-4h-oxazol-5-one, 2-oxazolin-5-one, 2-phenyl
PubChem CID 65073
ChEBI CHEBI:60296
IUPAC Name 2-phenyl-4H-1,3-oxazol-5-one
SMILES C1C(=O)OC(=N1)C2=CC=CC=C2

Specifications

Melting Point 89°C to 92°C
Quantity 25 g
Beilstein 128465
Sensitivity Air sensitive
Solubility Information Soluble in most common organic solvents.
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 2-Phenyl-5-oxazolone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.