missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylacetic dithioperoxyanhydride, 97%, BLD Pharm™

Product Code. 30783632
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
1g
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30783632 25 g 25g
30774409 5 g 5g
30766276 500 g 500g
30781826 100 g 100g
30774812 1 g 1g
30779817 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30783632 Supplier BLD Pharm Supplier No. BD14762425g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 15088-78-5
Molecular Formula C16H14O2S2
MDL Number MFCD00513572
Quantity 25 g
SMILES O=C(SSC(CC1=CC=CC=C1)=O)CC2=CC=CC=C2
Molecular Weight (g/mol) 302.41
Percent Purity 97%
Chemical Name or Material 2-Phenylacetic dithioperoxyanhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.