missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylacetimidamide Hydrochloride, TRC

Product Code. 30428345
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428345 250 mg -
30420460 1 g 1g
30412046 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428345 Supplier Toronto Research Chemicals Supplier No. TRCP335588250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2498-46-6
Molecular Formula C8H10N2 . HCl
Molecular Weight (g/mol) 170.64
Synonym Benzeneethanimidamide, hydrochloride (1:1) (ACI), Acetamidine, 2-phenyl-, hydrochloride (6CI, 7CI), Acetamidine, 2-phenyl-, monohydrochloride (8CI), Benzeneethanimidamide, monohydrochloride (9CI), 2-Phenylacetamidine hydrochloride, Phenylacetamidine hydrochloride, α-Phenylacetamidine hydrochloride, 2-Phenylacetimidamide hydrochloride
IUPAC Name 2-phenylethanimidamide;hydrochloride
SMILES Cl.NC(=N)Cc1ccccc1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C8H10N2.ClH/c9-8(10)6-7-4-2-1-3-5-7;/h1-5H,6H2,(H3,9,10);1H
Formula Weight 170.0611
Chemical Name or Material 2-Phenylacetimidamide Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.