missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylbenzimidazole-5-sulfonic acid, 98%

Product Code. 10259153 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10259153 250 g Glass bottle 250g
10611833 5 g Glass bottle 5g
10792471 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10259153 Supplier Thermo Scientific Acros Supplier No. 259702500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 27503-81-7 | C13H10N2O3S | 274.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27503-81-7
Molecular Formula C13H10N2O3S
Molecular Weight (g/mol) 274.29
MDL Number MFCD00053007
InChI Key UVCJGUGAGLDPAA-UHFFFAOYSA-N
Synonym ensulizole, 2-phenylbenzimidazole-5-sulfonic acid, phenylbenzimidazole sulfonic acid, 2-phenyl-1h-benzo d imidazole-5-sulfonic acid, 2-phenyl-1h-benzo d imidazole-6-sulfonic acid, 1h-benzimidazole-5-sulfonic acid, 2-phenyl, unii-9yq9di1w42, 2-phenyl-1h-benzimidazole-5-sulfonic acid, parsol hs, 2-phenyl-5-benzimidazolesulfonic acid
PubChem CID 33919
IUPAC Name 2-phenyl-3H-benzimidazole-5-sulfonic acid
SMILES OS(=O)(=O)C1=CC=C2N=C(NC2=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 250 g
Merck Index 15, 3647
Solubility Information Solubility in water: soluble
Formula Weight 274.28
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Phenylbenzimidazole-5-sulfonic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.