missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylbenzimidazole, 97%

Product Code. 11434443
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434443 5 g 5g
11444443 25 g 25g
11454443 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434443 Supplier Thermo Scientific Alfa Aesar Supplier No. A10996.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 716-79-0 | C13H10N2 | 194.24 g/mol

2-Phenylbenzimidazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenylbenzimidazole is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 716-79-0
Molecular Formula C13H10N2
Molecular Weight (g/mol) 194.24
MDL Number MFCD00005592
InChI Key DWYHDSLIWMUSOO-UHFFFAOYSA-N
Synonym 2-phenylbenzimidazole, 2-phenyl-1h-benzo d imidazole, phenizidole, phenzidole, phenzidol, gainex, 1h-benzimidazole, 2-phenyl, benzimidazole, 2-phenyl, 2-phenylbenzimdazole, 2-phenyl-1h-benzoimidazole
PubChem CID 12855
SMILES N1C2=CC=CC=C2N=C1C1=CC=CC=C1

Specifications

Melting Point 296°C to 297°C
Quantity 5 g
Beilstein 7087
Merck Index 14,7274
Solubility Information Slightly soluble in water.
IUPAC Name 2-phenyl-1H-1,3-benzodiazole
Formula Weight 194.24
Percent Purity 97%
Chemical Name or Material 2-Phenylbenzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.