missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Phenylbenzothiazole, 97%

Product Code. 11311567
Change view
Click to view available options
Quantity:
10g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11311567 10g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11311567 Supplier Alfa Aesar™ Supplier No. L02259.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 883-93-2
Boiling Point 371°C
Molecular Formula C13H9NS
MDL Number MFCD00005777
Quantity 10g
Beilstein 141340
Synonym 2-phenylbenzothiazole, 2-phenylbenzo d thiazole, benzothiazole, 2-phenyl, unii-fo573g4bgt, 2-phenyl benzothiazole, 2-phenylbenzthiazole, fo573g4bgt, wln: t56 bn dsj cr, phenyl-benzothiazole, benzothiazole,phenyl
InChI Key XBHOUXSGHYZCNH-UHFFFAOYSA-N
SMILES S1C2=CC=CC=C2N=C1C1=CC=CC=C1
IUPAC Name 2-phenyl-1,3-benzothiazole
Molecular Weight (g/mol) 211.28
PubChem CID 13439
Formula Weight 211.28
Percent Purity 97%
Chemical Name or Material 2-Phenylbenzothiazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.