missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethanol, 98+%

Product Code. 11452288
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452288 5000 g 5000g
11442288 250 g 250g
11432288 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452288 Supplier Thermo Scientific Alfa Aesar Supplier No. A15241.0I
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 60-12-8 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Phenylethanol is used also as an aromatic essence and preservative in pharmaceutics and perfumery. It is also used as odor agents, air care products, cleaning and furnishing care products, laundry and dishwashing products, personal care products.

Solubility
Miscible with chloroform.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-12-8
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002886
InChI Key WRMNZCZEMHIOCP-UHFFFAOYSA-N
Synonym phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol
PubChem CID 6054
ChEBI CHEBI:49000
SMILES OCCC1=CC=CC=C1

Specifications

Melting Point -27°C
Density 1.021
Boiling Point 219°C
Flash Point 101°C (214°F)
Assay Percent Range ≥98%
Linear Formula C6H5(CH2)2OH
Refractive Index 1.533
Quantity 5000 g
UN Number UN2810
Beilstein 1905732
Merck Index 14,7224
Solubility Information Miscible with chloroform.
IUPAC Name 2-phenylethan-1-ol
Formula Weight 122.17
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 2-Phenylethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.