missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethanol, >99%, MP Biomedicals™

Product Code. 11423542 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423542 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11423542 Supplier MP Biomedicals Supplier No. 0215617691
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 60-12-8
Color Colorless
Density 1.02g/cm3 at 20°C
Molecular Formula C8H10O
MDL Number MFCD00002886
Quantity 1 L
Synonym phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol
InChI Key WRMNZCZEMHIOCP-UHFFFAOYSA-N
SMILES OCCC1=CC=CC=C1
IUPAC Name 2-phenylethan-1-ol
Molecular Weight (g/mol) 122.17
PubChem CID 6054
ChEBI CHEBI:49000
Formula Weight 122.3
Percent Purity >99%
Physical Form Slightly Viscous Liquid
Chemical Name or Material 2-Phenylethyl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.