missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylethyl-1,1,2,2-d4-amine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Phenethylamine-d4 |
| CAS | 87620-08-4 |
| Molecular Formula | C8 2H4 H7 N |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C8H11N/c9-7-6-8-4-2-1-3-5-8/h1-5 H,6-7,9H2/i6D2,7D2 |
| SMILES | [2 H]C([2 H])(N)C([2 H])([2 H])c1ccccc1 |
| IUPAC Name | 1,1,2,2-tetradeuterio-2-phenylethanamine |
| Molecular Weight (g/mol) | 125.2 |
| Formula Weight | 125.1143 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction