missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethyl acetate, 98%

Product Code. 11444757
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Dimensione della confezione
11444757 250 g 250g
11434757 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11444757 Fornitore Thermo Scientific Alfa Aesar N. del fornitore B21238.30
 Restricted Item
Estimated Shipment: 03-12-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 103-45-7 | C10H12O2 | 164.20 g/mol

Phenylethyl acetate mantains the typical rose notes of Phenylethanol with a more intense honey profile which is perfect for fruit and alcoholic drink applications. It is used as a pharmaceutical intermediate and also it holds application in gas chromatography and liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylethyl acetate mantains the typical rose notes of Phenylethanol with a more intense honey profile which is perfect for fruit and alcoholic drink applications. It is used as a pharmaceutical intermediate and also it holds application in gas chromatography and liquid chromatography.

Solubility
It is insoluble in paraffin oil, water. Soluble in alcohol, dipropylene glycol, fixed oils, kerosene, propylene glycol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Protected from heat and light.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 103-45-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00008720
InChI Key MDHYEMXUFSJLGV-UHFFFAOYSA-N
Synonym phenethyl acetate, 2-phenethyl acetate, acetic acid, 2-phenylethyl ester, benzylcarbinyl acetate, beta-phenylethyl acetate, acetic acid, phenethyl ester, phenethyl alcohol, acetate, phenylethyl acetate, acetic acid phenethyl ester, ethanol, 2-phenyl-, acetate
PubChem CID 7654
ChEBI CHEBI:31988
IUPAC Name 2-phenylethyl acetate
SMILES CC(=O)OCCC1=CC=CC=C1

Specifica

Melting Point -31°C
Density 1.029
Boiling Point 231°C to 233°C
Flash Point 107°C (225°F)
Odor Fruit-like
Refractive Index 1.499
Quantity 250 g
Beilstein 638179
Solubility Information It is insoluble in paraffin oil,water. Soluble in alcohol,dipropylene glycol,fixed oils,kerosene,propylene glycol.
Formula Weight 164.21
Percent Purity 98%
Chemical Name or Material 2-Phenylethyl acetate
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.