missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethyl isothiocyanate, 98%

Product Code. 11432088
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432088 25 g 25g
11422088 5 g 5g
11442088 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11432088 Supplier Thermo Scientific Alfa Aesar Supplier No. A15123.14
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2257-09-2 | C9H9NS | 163.24 g/mol

Phenethyl isothiocyanate is an NNK-induced lung tumorigenesis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenethyl isothiocyanate is an NNK-induced lung tumorigenesis inhibitor.

Solubility
Soluble in chloroform, ethyl acetate, and dichoromethane. Reacts with water.

Notes
Moisture Sensitive, store away from moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agent. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2257-09-2
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.24
MDL Number MFCD00004821
InChI Key IZJDOKYDEWTZSO-UHFFFAOYSA-N
Synonym 2-phenylethyl isothiocyanate, phenethyl isothiocyanate, phenylethyl isothiocyanate, 2-isothiocyanatoethyl benzene, phenethyl mustard oil, phenylaethylsenfoel, benzene, 2-isothiocyanatoethyl, peitc, phenylethyl mustard oil, 2-phenylethylisothiocyanate
PubChem CID 16741
ChEBI CHEBI:351346
IUPAC Name 2-isothiocyanatoethylbenzene
SMILES C1=CC=C(C=C1)CCN=C=S

Specifications

Density 1.09
Boiling Point 139°C to 140°C (11 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.5888
Quantity 25 g
UN Number UN2922
Beilstein 2084162
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform,ethyl acetate,and dichoromethane. Reacts with water.
Formula Weight 163.24
Percent Purity 98%
Chemical Name or Material 2-Phenylethyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.