missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylindole, 95%

Product Code. 11410088
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410088 250 g 250g
11400088 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410088 Supplier Thermo Scientific Alfa Aesar Supplier No. B23674.30
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 948-65-2 | C14H11N | 193.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 948-65-2
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD00005608
InChI Key KLLLJCACIRKBDT-UHFFFAOYSA-N
Synonym 2-phenylindole, 1h-indole, 2-phenyl, indole, 2-phenyl, stabilizer i, alpha-phenylindole, 2-phenyl indole, phenylindole, unii-mqd44hv3p1, .alpha.-phenylindole, mqd44hv3p1
PubChem CID 13698
IUPAC Name 2-phenyl-1H-indole
SMILES C1=CC=C(C=C1)C2=CC3=CC=CC=C3N2

Specifications

Melting Point 189°C to 190°C
Boiling Point 250°C (10 mmHg)
Quantity 250 g
Formula Weight 193.25
Percent Purity 95%
Chemical Name or Material 2-Phenylindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.