missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpentane, tech. 85%

Product Code. 11460631
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460631 5 g 5g
11369559 1 g 1g
11470631 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460631 Supplier Thermo Scientific Alfa Aesar Supplier No. B20114.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2719-52-0 | C11H16 | 148.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2719-52-0
Molecular Formula C11H16
Molecular Weight (g/mol) 148.25
MDL Number MFCD00027145
InChI Key LTHAIAJHDPJXLG-UHFFFAOYNA-N
Synonym 2-phenylpentane, sec-pentylbenzene, benzene, sec-pentyl, sec-amylbenzene, benzene, 1-methylbutyl, 1-phenyl-1-methylbutane, s-amylbenzene, d-2-phenyl-pentane, sec-amyl benzene, methylbutyl benzene
PubChem CID 17627
IUPAC Name pentan-2-ylbenzene
SMILES CCCC(C)C1=CC=CC=C1

Specifications

Density 0.86
Quantity 5 g
Formula Weight 148.25
Percent Purity 85%
Grade Technical
Chemical Name or Material 2-Phenylpentane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.