missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylphenol, TRC

Product Code. 30010774
Change view
Click to view available options
Quantity:
1 g
10 g
500 g
Unit Size:
10g
1g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010774 10 g 10g
30007981 500 g 500g
17971405 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010774 Supplier Toronto Research Chemicals Supplier No. TRCP33587010G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-43-7
Molecular Formula C12 H10 O
Molecular Weight (g/mol) 170.21
Synonym [1,1'-Biphenyl]-2-ol, 2-Biphenylol (8CI), 2-Hydroxy-1,1'-biphenyl, 2-Hydroxybiphenyl, 2-Hydroxydiphenyl, 2-Phenylphenol, Anthrapole 73, Britewood S, Decco OPP 20, Dowicide 1, Invalon OP, NSC 1548, Nectryl, O-PP, Preventol 3041, Preventol O extra, Remol TRF, Tetrosin OE, Tetrosin OE-N, Tumescal OPE, o-Biphenylol, o-Diphenylol, o-Hydroxybiphenyl, o-Hydroxydiphenyl, o-Phenylphenol, o-Xenol, E231
IUPAC Name 2-phenylphenol
SMILES OC=1C=CC=CC1C=2C=CC=CC2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H10O/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9,13H
Formula Weight 170.0732
Chemical Name or Material 2-Phenylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.