missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpiperazine, 96%

Product Code. 11495880
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495880 25 g 25g
11485880 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495880 Supplier Thermo Scientific Alfa Aesar Supplier No. H26502.14
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5271-26-1 | C10H14N2 | 162.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5271-26-1
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.236
MDL Number MFCD01871362
InChI Key RIMRLBGNCLMSNH-UHFFFAOYSA-N
Synonym 2-phenyl-piperazine, piperazine, 2-phenyl, 2-phenylpiperazin, 3-phenylpiperazine, 2-phenyl piperazine, pubchem8559, acmc-20a0jb, rs-2-phenylpiperazine, 2-phenylpiperazine, +,--2-phenyl-piperazine
PubChem CID 250673
IUPAC Name 2-phenylpiperazine
SMILES C1CNC(CN1)C2=CC=CC=C2

Specifications

Melting Point 83°C to 87°C
Quantity 25 g
UN Number UN3259
Sensitivity Air Sensitive
Formula Weight 162.24
Percent Purity 96%
Chemical Name or Material 2-Phenylpiperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.