missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylpropane-1,1,1,2,3,3,3-d7, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Isopropylbenzene-D7 (isopropyl-D7) |
| CAS | 20201-28-9 |
| Molecular Formula | C9 D7 H5 |
| Quantity | 0.5 g |
| Synonym | Benzene, [1-(methyl-d3)ethyl-1,2,2,2-d4]- (9 CI), Cumene-α,β,β,β,β',β',β'-d7 (8 CI), 1,1,1,2,3,3,3-Heptadeuteriopropan-2-ylbenzene, Dimethylphenylmethane-d7, [1-(Methyl-d3)ethyl-1,2,2,2-d4]benzene, Isopropylbenzene-d7, Isopropylbenzene-D7 (isopropyl-D7), 2-Phenylpropane-1,1,1,2,3,3,3-d7 |
| InChI Formula | InChI=1 S/C9H12/c1-8(2)9-6-4-3-5-7-9/h3-8 H,1-2H3/i1D3,2D3,8 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(c1ccccc1)C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,2,3,3,3-heptadeuteriopropan-2-ylbenzene |
| Molecular Weight (g/mol) | 127.2347 |
| Formula Weight | 127.1378 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction