missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpropionic Acid Ethyl Ester, TRC

Product Code. 30426746
Change view
Click to view available options
Quantity:
25 mg
250 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426746 500 mg 500mg
30412042 250 mg -
30417577 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426746 Supplier Toronto Research Chemicals Supplier No. TRCP336180500MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2510-99-8
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
Synonym Ethyl 2-Phenylpropanoate, Ethyl 2-Phenylpropionate, Ethyl Hydrotropate, Ethyl alpha-Phenylpropionate, NSC 254979, alpha-Methylbenzeneacetic Acid Ethyl Ester
IUPAC Name ethyl 2-phenylpropanoate
SMILES CCOC(=O)C(C)c1ccccc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H14O2/c1-3-13-11(12)9(2)10-7-5-4-6-8-10/h4-9H,3H2,1-2H3
Formula Weight 178.0994
Chemical Name or Material 2-Phenylpropionic Acid Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.