missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpyridine, 97%

Product Code. 10351424 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10351424 5 g Glass bottle 5g
10606744 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10351424 Supplier Thermo Scientific Acros Supplier No. 130900050
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 1008-89-5 | C11H9N | 155.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1008-89-5
Molecular Formula C11H9N
Molecular Weight (g/mol) 155.20
MDL Number MFCD00006280,MFCD31699954
InChI Key VQGHOUODWALEFC-UHFFFAOYSA-N
Synonym pyridine, 2-phenyl, o-phenylpyridine, pyridine, phenyl, 2-phenyl-pyridine, phenyl pyridine, 2-azabiphenyl, 2-phenyl pyridine, 2-phenylpyridine, pubchem13085, 2-phenylpyridine????
PubChem CID 13887
IUPAC Name 2-phenylpyridine
SMILES C1=CC=C(C=C1)C1=CC=CC=N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0860g/mL
Boiling Point 268°C to 270°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)C6H5
Refractive Index 1.6220 to 1.624
Quantity 5 g
Beilstein 20,424
Specific Gravity 1.086
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible in ethanol,ethyl ether
Formula Weight 155.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Phenylpyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Phenylpyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.