missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpyridine, 98%

Product Code. 11392077
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392077 10 g 10g
11302087 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392077 Supplier Thermo Scientific Alfa Aesar Supplier No. L07861.09
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1008-89-5 | C11H9N | 155.20 g/mol

2-Phenylpyridine is used to produce 2-phenyl-pyridine-1-oxide. It is used in the manufacturing of organic light emitting diodes (OLEDs). It is used as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenylpyridine is used to produce 2-phenyl-pyridine-1-oxide. It is used in the manufacturing of organic light emitting diodes (OLEDs). It is used as an intermediate.

Solubility
Fully miscible with ethanol, acetone, toluene, methylene chloride. Not miscible in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1008-89-5
Molecular Formula C11H9N
Molecular Weight (g/mol) 155.20
MDL Number MFCD00006280,MFCD31699954
InChI Key VQGHOUODWALEFC-UHFFFAOYSA-N
Synonym pyridine, 2-phenyl, o-phenylpyridine, pyridine, phenyl, 2-phenyl-pyridine, phenyl pyridine, 2-azabiphenyl, 2-phenyl pyridine, 2-phenylpyridine, pubchem13085, 2-phenylpyridine????
PubChem CID 13887
IUPAC Name 2-phenylpyridine
SMILES C1=CC=C(C=C1)C1=CC=CC=N1

Specifications

Melting Point -5°C
Density 1.086
Boiling Point 268°C to 270°C
Flash Point >110°C (230°F)
Refractive Index 1.624
Quantity 10 g
Beilstein 110445
Solubility Information Fully miscible with ethanol,acetone,toluene,methylene chloride. Not miscible in water.
Formula Weight 155.2
Percent Purity 98%
Chemical Name or Material 2-Phenylpyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.