missing translation for 'onlineSavingsMsg'
Learn More

2-(Phenylthio)ethanethioamide, 97%, Thermo Scientific™

Product Code. 10523212
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
Product Code. Quantity unitSize
10523212 1 g 1g
10435702 10 g 10g
10158224 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 10523212

Brand: Thermo Scientific Maybridge KM04192DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 59865-82-6
Molecular Formula C8H9NS2
Molecular Weight (g/mol) 183.29
MDL Number MFCD00085154
InChI Key ZRDOSZLCJHRADP-UHFFFAOYSA-N
Synonym 2-phenylthio ethanethioamide, 2-phenylsulfanyl ethanethioamide, maybridge1_004803, 2-phenylthio thioacetamide, 2-phenylsulfanyl-thioacetamide, ethanethioamide,2-phenylthio, 1-amino-2-phenylthioethane-1-thione
PubChem CID 2778855
SMILES NC(=S)CSC1=CC=CC=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Linear Formula C6H5SCH2CSNH2
Quantity 1 g
IUPAC Name 2-phenylsulfanylethanethioamide
Formula Weight 183.3
Percent Purity 95%
Chemical Name or Material 2-(Phenylthio)ethanethioamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.