missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylthioacetamide, 97%

Product Code. 15413487
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15413487 250 mg 250mg
11435910 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15413487 Supplier Thermo Scientific Alfa Aesar Supplier No. H26514.MD
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 645-54-5 | C8H9NS | 151.227 g/mol

2-Phenylthioacetamide find extensive use in organic synthesis. N-bromo and N-chloro succinimides are halogenating agents. Phthalimides are used in the synthesis of primary amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenylthioacetamide find extensive use in organic synthesis. N-bromo and N-chloro succinimides are halogenating agents. Phthalimides are used in the synthesis of primary amines.

Solubility
Slightly Soluble in water (1.8 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-54-5
Molecular Formula C8H9NS
Molecular Weight (g/mol) 151.227
MDL Number MFCD00022177
InChI Key CJXBHFANXQMZBF-UHFFFAOYSA-N
Synonym 2-phenylthioacetamide, benzeneethanethioamide, phenylthioacetamide, acetamide, 2-phenylthio, benzeneethanimidothioic acid, phenylacetothioamide, 1-amino-2-phenylethane-1-thione, a-phenylthioacetamide, 2-phenyl-thioacetamide, alpha-phenylthioacetamide
PubChem CID 731368
IUPAC Name 2-phenylethanethioamide
SMILES C1=CC=C(C=C1)CC(=S)N

Specifications

Melting Point 91°C to 95°C
Quantity 250 mg
Solubility Information Slightly Soluble in water (1.8g/L) (25°C).
Formula Weight 151.23
Percent Purity 97%
Chemical Name or Material 2-Phenylthioacetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.