missing translation for 'onlineSavingsMsg'
Learn More

2-Phospho-L-ascorbic Acid Trisodium Salt, TRC

Product Code. 30414180
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414180 25 g -
30410268 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30414180 Supplier Toronto Research Chemicals Supplier No. TRCP35400325G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66170-10-3
Molecular Formula C6H6Na3O9P
Molecular Weight (g/mol) 322.05
Synonym Ascorbic Acid Phosphate Sodium Salt, L-Ascorbic Acid 2-(Dihydrogen Phosphate) Trisodium Salt, Sodium Ascorbyl Phosphate, Stay-C 50, Trisodium Ascorbate-2-phosphate, VCP-NA
IUPAC Name trisodium;[(2R)-2-[(1S)-1,2-dihydroxyethyl]-3-oxido-5-oxo-2H-furan-4-yl] phosphate
SMILES [Na+].[Na+].[Na+].OC[C@H](O)[C@H]1OC(=O)C(=C1[O-])OP(=O)([O-])[O-]

Specifications

Quantity 25 g
InChI Formula InChI=1S/C6H9O9P.3Na/c7-1-2(8)4-3(9)5(6(10)14-4)15-16(11,12)13;;;/h2,4,7-9H,1H2,(H2,11,12,13);;;/q;3*+1/p-3/t2-,4+;;;/m0.../s1
Formula Weight 321.9443
Chemical Name or Material 2-Phospho-L-ascorbic Acid Trisodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.